Vortex-Genie 2T Shakers
Enhanced vortex action for maximum high-speed touch mixing
  • Solid metal casing with rubber feet prevents shaker from ''walking'' across your benchtop
Vortex-Genie® 2T Shaker features both touch and hands-free operation. Shake samples from 1 to 60 seconds in touch mode, 1 to 60 minutes in hands-free mode, or continuously in the automatic untimed mode. The 2T shaker accepts all standard Genie attachments and accessories making it an economical addition to you laboratory. Includes pop-off cup and 3'' platform.
123,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
Scientific Industries Vortex-Genie SI-T236 2T Shaker, 600 to 3200 rpm, 120 VAC WW-04724-20 Mfr # SI-T236
COMPARE
120
₹123,000.00INR / Each
Scientific Industries Vortex-Genie SI-T246 2T Shaker, 600 to 2700 rpm, 230 VAC WW-04724-25 Mfr # SI-T246
COMPARE
230
₹123,000.00INR / Each