BriskHeat® Very High Temperature Heating Tapes for Non-Conductive Surfaces-240 VAC
Flexible, Multi-purpose direct-contact heaters for use for on non-conductive surfaces such as glassware
  • Maximum exposure temperature of 1400°F (760°C)
  • Standard-duty insulated design
  • Nongrounded
Resistance wires are double insulated with braided SAMOX yarns, then knit into flat tapes. .
The 240 VAC tapes have bare leads with opposite-end configuration and are supplied with ties at each end.
Warning: Heating tapes should not be overlapped and require the use of a temperature or power controller sold separately
12,900.00 - ₹16,500.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat TBW0-102-040 Very High Temperature (1400C) Heating Tape, 48" L x 1" W, 240V/620W WW-36119-38 Mfr # TBW0-102-040
COMPARE
48 1400 240
₹16,500.00INR / Each
BriskHeat TBW0-052-020 Very High Temperature (1400C) Heating Tape, 24" L x 1/2" W, 240V/160W WW-36119-41 Mfr # TBW0-052-020
COMPARE
24 1400 240
₹12,900.00INR / Each