BriskHeat® Very High Temperature Heating Tapes for Non-Conductive Surfaces-120 VAC

Flexible, Multi-purpose direct-contact heaters for use for on non-conductive surfaces such as glassware
  • Maximum exposure temperature of 1400°F (760°C)
  • Standard-duty insulated design
  • Nongrounded
Resistance wires are double insulated with braided SAMOX yarns, then knit into flat tapes.

Tapes include ties at each end, anda standard two-prong split plug

Warning: Heating tapes should not be overlapped and require the use of a temperature or power controller sold separately
12,000.00 - ₹20,500.00INR / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Max Temperature (° F) Power (VAC)
Availability Pricing
BriskHeat TBW0-051-020 Insulated Heating Tape, 24" L x 1/2" W x 120V; 160 Watt WW-36119-30 Mfr # TBW0-051-020
COMPARE
24 1400 120
₹12,000.00INR / Each
BriskHeat TBW0-051-040 Insulated Heating Tape, 48" L x 1/2" W x 120V; 310 Watt WW-36119-31 Mfr # TBW0-051-040
COMPARE
48 1400 120
₹14,000.00INR / Each
BriskHeat TBW0-051-060 Insulated Heating Tape, 72" L x 1/2" W x 120V; 470 Watt WW-36119-32 Mfr # TBW0-051-060
COMPARE
72 1400 120
₹17,600.00INR / Each
BriskHeat TBW0-051-080 Insulated Heating Tape, 92" L x 1/2" W x 120V; 620 Watt WW-36119-33 Mfr # TBW0-051-080
COMPARE
96 1400 120
₹19,100.00INR / Each
BriskHeat TBW0-101-060 Insulated Heating Tape, 72" L x 1" W x 120V; 940 Watt WW-36119-36 Mfr # TBW0-101-060
COMPARE
72 1400 120
₹20,500.00INR / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat