Wildco® Vertical Alpha Water Samplers
Suitable for use in oceans, deep lakes, and corrosive waters
  • Urethane end seals snap shut with minimum surface disturbance on messenger contact
  • Drain valve provides easy sample removal
Vertical samplers retrieve large water samples at any depth and collect plankton or floating sediments. Samplers include a carrying case; order messenger and line separately. Silicone end seals are available by special order.
152,000.00 - ₹167,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material
Availability Pricing
Wildco 1120-D40 Vertical alpha water sampler, PVC, 2.2 L WW-05487-00 Mfr # 1120-D40
COMPARE
PVC
₹152,000.00INR / Each
Wildco 1120-C40 Vertical alpha water sampler, acrylic, 2.2 L WW-05487-10 Mfr # 1120-C40
COMPARE
Acrylic
₹167,000.00INR / Each