Velp Digital Ceramic Stirring Hot Plates
Top plate and body resist acids, bases and solvents
  • Controls and top plate are separated to increase safety
  • Digital “HOT PLATE” warning signal when the plate temperature is above 50°C
Easy-to-clean, white ceramic top is highly resistant to corrosion and is excellent for observing liquid color changes. Built-in electronics keep stirring speeds constant during heavy loads or changing viscosities. Microprocessor control allows for precise temperature and speed control. Control panel is separated from the main body, protecting electronics from splashes and spills. Stirrer features a “HOT PLATE” warning signal that remains illuminated while the temperature is on and will flash if the instrument is turned off until plate temperature drops below 50°C.
64,000.00 - ₹82,600.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Description
Availability Pricing
Velp 20510011 Digital Ceramic Stirring Hot Plate, 120V WW-86579-44 Mfr # F20510551
COMPARE
115 Digital Ceramic Stirring Hot Plate, 120V
₹64,000.00INR / Each
Velp 20500011 Digital Ceramic Stirring Hot Plate, 240V WW-86579-45 Mfr # F20500551
COMPARE
230 Digital Ceramic Stirring Hot Plate, 240V
₹82,600.00INR / Each
Velp 20510061 Digital Ceramic Stirring Hot Plate Thermoregulated, 120V WW-86579-46 Mfr # F20510554
COMPARE
115 Digital Ceramic Stirring Hot Plate Thermoregulated, 120V
₹74,500.00INR / Each
Velp 20510051 Digital Ceramic Stirring Hot Plate, with Timer, 120V WW-86579-48 Mfr # SA20510550
COMPARE
115 Digital Ceramic Stirring Hot Plate, with Timer, 120V
₹75,600.00INR / Each