This product is now discontinued, View Related Products
Vector Laboratories WestVision™ Peroxidase Polymers for Western Blot Detection
Improve your signal intensity without increasing background
  • Save time with ready-to-use format
WestVision™ Block and Diluent is a ready-to-use Tris buffer-based blocking solution intended for Western or dot blot applications. This solution is a proprietary formulation containing Tween 20 that improves signal intensity and resolution without increasing background. To achieve the best results, use WestVision™ Block and Diluent for all blocking and diluting steps including primary antibody and secondary antibody detection reagents. WestVision™ Block and Diluent is compatible with both alkaline phosphatase- and peroxidase- based detections systems, as well as with both chemiluminescence and chromogenic development.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Vector Laboratories WestVision Block and Diluent for Western Blots, 500 ml WW-93952-10 Mfr # SP-7000-500
COMPARE
Block and Diluent for Western Blots, 500 ml
Call For Price
Vector Laboratories WestVision HRP Anti-Mouse IgG for Western Blot Detection WW-93952-11 Mfr # WB-2000-.8
COMPARE
HRP Anti-Mouse IgG for Western Blot Detection