This product is now discontinued, View Related Products
Vector Laboratories DuoLuX™ Chemiluminescent / Fluorescent Substrate
Blots luminesce and fluoresce for easy visualization
  • Strong luminescent signal lasts up to 8 hours
  • Fluorescence is permanent and detectable for months
DuoLux substrate kits are available for both peroxidase and alkaline phosphatase reactions to produce a strong chemiluminescent and fluorescent signal. The substrate luminesces in the blue range, with a peak emission at 453 nm. Fluorescent excitation occurs over a range of wavelengths: 254 nm, 365 nm, with maximum excitation at 405 nm. Maximum fluorescent emission occurs at 453 nm.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Vector Laboratories DuoLux Chemiluminescent/Fluorescent Substrate for Peroxidase, 200 ml WW-93952-08 Mfr # SK-6604-200
COMPARE
Chemiluminescent/Fluorescent Substrate for Peroxidase, 200 ml
Call For Price
Vector Laboratories DuoLux Chemiluminescent/Fluorescent Substrate for Alkaline Phosphatase, 100 ml WW-93952-09 Mfr # SK-6605-100
COMPARE
Chemiluminescent/Fluorescent Substrate for Alkaline Phosphatase, 100 ml