This product is now discontinued, View Related Products
Value Plastics RQ Series Quick Disconnects for Bioprocessing
Guard against Disconnects with a Secure Lock
  • Materials meet USP Class VI and ISO 10993 criteria
  • Several connector configurations available in commonly used polypropylene and polycarbonate materials
  • Large ergonomic button and thumb pad with secure latch and optional integrated locking mechanism
  • Plugs and caps available to maintain fluid path integrity
  • RQC Tubing Sizes:
  • • 1/4" (6.4 mm) and 3/8" (9.5 mm) tubing IDs
  • RQX Tubing Sizes:
  • • 3/8" (9.5 mm) and 1/2" (12.7 mm) tubing IDs
RQ Series offers a cost effective solution for flexible tubing connections in single use systems. The connector set features a user-friendly interface that includes a large guarded button for a secure lock to guard against disconnects.
All materials satisfy USP Class VI and ISO 10993 criteria and are suitable for autoclave and EtO or gamma sterilization.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Connection Type Description
Availability Pricing
Value Plastics RQC Series Female Open Flow Connector 600 Series Barb 1/4"; (6.4 mm) ID Tubing Polycarbonate 100 WW-15957-11 Mfr # RQCF655-9024-001
COMPARE
Quick-Disconnect x Barb RQC Series Female Open Flow Connector 600 Series Barb 1/4\"; (6.4 mm) ID Tubing Polycarbonate 100
Call For Price
Value Plastics RQC Series Female Connector Cap Polycarbonate 100/pack WW-15957-19 Mfr # RQCFC-9024-001
COMPARE
Quick-Disconnect x Barb RQC Series Female Connector Cap Polycarbonate 100/pack
Call For Price
Value Plastics RQC Series Male Open Flow Connector 600 Series Barb 1/4"; (6.4 mm) ID Tubing Polycarbonate Sili WW-15957-23 Mfr # RQCM655-9024-001
COMPARE
Quick-Disconnect x Barb RQC Series Male Open Flow Connector 600 Series Barb 1/4\"; (6.4 mm) ID Tubing Polycarbonate Sili
Call For Price
Value Plastics RQC Series Male Connector Plug Polycarbonate Silicone O-Ring 100/pack WW-15957-27 Mfr # RQCMP-9024-001
COMPARE
Quick-Disconnect x Barb RQC Series Male Connector Plug Polycarbonate Silicone O-Ring 100/pack