This product is now discontinued, View Related Products
Uvex by Honeywell SVP-Series Safety Glasses
Premium look, safety, and comfort at an economic price
  • Choose the frame color to suit your tastes Cushioned nose and ear grips for superior comfort and stay Clear polycarbonate lenses are ANSI Z.81.1+2015 certified Select from Hardcoat/anti-scratch or Uvextra Anti-Fog lens coatings 200-series features thin frame vs 400-series thicker frame
3,400.00 - ₹4,400.00INR / Box of 10
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Frame Color Lens Coating Description
Availability Pricing
Uvex by Honeywell SVP200 Safety Glasses, Clear Frame, Clear Hardcoat Lens; 10/Bx; 10/Bx WW-86497-71 Mfr # SVP200
COMPARE
Clear Hardcoat anti-scratch Safety Glasses, Clear Frame, Clear Hardcoat Lens; 10/Bx
₹3,600.00INR / Box of 10
Uvex by Honeywell SVP201 Safety Glasses, Clear Frame, Clear Uvextra Anti-Fog Lens; 10/Bx WW-86497-72 Mfr # SVP201
COMPARE
Clear Uvextra anti-fog Safety Glasses, Clear Frame, Clear Uvextra Anti-Fog Lens; 10/Bx
₹3,400.00INR / Box of 10
Uvex by Honeywell SVP205 Safety Glasses, Blue Frame, Clear Uvextra Anti-Fog Lens; 10/Bx WW-86497-73 Mfr # SVP205
COMPARE
Blue Uvextra anti-fog Safety Glasses, Blue Frame, Clear Uvextra Anti-Fog Lens; 10/Bx
₹3,700.00INR / Box of 10
Uvex by Honeywell SVP207 Safety Glasses, Blue Frame, Hardcoat Lens; 10/Bx WW-86497-74 Mfr # SVP207
COMPARE
Blue Hardcoat anti-scratch Safety Glasses, Blue Frame, Hardcoat Lens ; 10/Bx
Call For Price
Uvex by Honeywell SVP208 Safety Glasses, Pink Frame, Clear Uvextra Anti-Fog Lens; 10/Bx WW-86497-75 Mfr # SVP208
COMPARE
Pink Uvextra anti-fog Safety Glasses, Pink Frame, Clear Uvextra Anti-Fog Lens; 10/Bx
₹3,700.00INR / Box of 10
Uvex by Honeywell SVP400 Safety Glasses, Clear Frame, Clear Hardcoat Lens; 10/Bx WW-86497-76 Mfr # SVP400
COMPARE
Clear Hardcoat anti-scratch Safety Glasses, Clear Frame, Clear Hardcoat Lens; 10/Bx
₹4,400.00INR / Box of 10
Uvex by Honeywell SVP401 Safety Glasses, Clear Frame, Clear Uvextra Anti-Fog Lens; 10/Bx WW-86497-77 Mfr # SVP401
COMPARE
Clear Uvextra anti-fog Safety Glasses, Clear Frame, Clear Uvextra Anti-Fog Lens; 10/Bx
Call For Price
Uvex by Honeywell SVP404 Safety Glasses, Gray Frame, Hardcoat Lens; 10/Bx WW-86497-78 Mfr # SVP404
COMPARE
Gray Hardcoat anti-scratch Safety Glasses, Gray Frame, Hardcoat Lens; 10/Bx
Call For Price
Showing 1- 8 of 8