This product is now discontinued, View Related Products
Uvex by Honeywell Hypershock™ Safety Glasses
High-performance wraparound eyewear with a sporty style
  • Protects against impact from sun, wind and dust, as well as airborne debris HydroShield long-life antifog coating to help keep lenses clear Soft molded inserts and nosepiece provide a comfortable, secure fit
Sharp and modern looking, these clear safety glasses have superior side protection and peripheral vision. Hypershock glasses are 100-percent dielectric so, you can wear them in environments where accidental exposure to an electrical charge is possible. Polycarbonate lens features antiscratch and HydroShield antifog hard coatings.
Compliance: ANSI Z87.1-2015, CSA Z94.3
22,600.00INR / Box of 10
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Frame Color Lens Coating Lens Color
Availability Pricing
Uvex by Honeywell S2960HS Hypershock Safety Glasses, Brown Frame, Clear, HydroShield Antifog Lens; 10/BX WW-86497-97 Mfr # S2960HS
COMPARE
Brown Antifog Clear
₹22,600.00INR / Box of 10