This product is now discontinued, View Related Products
UV Translinker
UV crosslinker and transilluminator in one self-contained space-saving unit
  • Crosslinker uses 254 nm UV tube which provides uniform radiation for crosslinking DNA or RNA to membranes
  • Transilluminator uses 302 nm UV tube in Hi/Lo intensity styles
The Translinker has all of the features of the Crosslinker and utilizes shortwave UV in the Crosslinker chamber. On/Off controls for the transiluminator are included on the Crosslinker panel. Filter surface measures 20 x 20cm. The 115V model is ETL/ETLc certified, and the 230V model is CE certified. Transilluminator portion carries a 2-year warranty while crosslinker portion carries a 1-year warranty.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (watts)
Availability Pricing
UV Translinker, ETL/Ecertified, 115V/60Hz WW-09814-43 Mfr # 95-0300-01
COMPARE
Call For Price
UV Translinker, ETL/Ecertified, 230V/50Hz WW-09814-45 Mfr # 95-0300-02