US Industrial Analog Gas Detector / Alarm
Don’t risk workers’ safety
  • Continuous 24 hour monitoring Compact, durable units Explosion proof housing for sensor Audio/visual alarms Malfunction LED and test/checking switches Infrequent maintenance and calibration required Remote sensor reliable to 600 feet from controller Relay contacts and recorder terminals
The wall mount unit is for use in an enclosed area. Remote sensor provides open space monitoring ability. At hazardous locations, explosion proof sensors and their junction box housing meet NEC requirements for Class 1, Division 1, Group A-B-C-D. The unit is designed for those who are concerned about explosive and toxic gases over a long period of time mostly within an enclosed area and/or work environment.
Call For Price
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Gases Measured
Availability Pricing
US Industrial 6200R Analog Gas Detector/Alarm, LP Gas/Gasoline Vapor, Wall Mount with remote sensor WW-10110-19 Mfr # 6200R
COMPARE
Liquid propane gas/gasoline vapor