This product is now discontinued, View Related Products
Koflo Unmounted In-Line Static Mixers
Works well with flexible tubing
Unmounted mixers can be used with any standard tubing having the same tubing ID as the element OD given in the table. They work well with flexible tubing.
7,200.00 - ₹14,100.00INR / Pkg of 10
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Mixer Length (in) No. Of Elements Description
Availability Pricing
Koflo 3/16-12P Unmounted in-line mixer; 3/16"OD, 12 elements, 10/pack WW-04667-02 Mfr # 3/16-12P
COMPARE
2.375 12 Unmounted in-line mixer; 3/16\"OD, 12 elements, 10/pack
₹7,200.00INR / Pkg of 10
Koflo 1/4-12P Unmounted in-line mixer; 1/4"OD, 12 elements, 10/pack WW-04667-04 Mfr # 1/4-12P
COMPARE
3 12 Unmounted in-line mixer; 1/4\"OD, 12 elements, 10/pack
₹7,500.00INR / Pkg of 10
Koflo 3/8-12P Unmounted in-line mixer; 3/8"OD, 12 elements, 10/pack WW-04667-06 Mfr # 3/8-12P
COMPARE
4.75 12 Unmounted in-line mixer; 3/8\"OD, 12 elements, 10/pack
₹7,900.00INR / Pkg of 10
Koflo 1/2-12P Unmounted in-line mixer; 1/2"OD, 12 elements, 10/pack WW-04667-08 Mfr # 1/2-12P
COMPARE
5 12 Unmounted in-line mixer; 1/2\"OD, 12 elements, 10/pack
₹11,200.00INR / Pkg of 10
Koflo 3/16-24P Unmounted in-line mixer; 3/16"OD, 24 elements, 10/pack WW-04667-12 Mfr # 3/16-24P
COMPARE
4.625 24 Unmounted in-line mixer; 3/16\"OD, 24 elements, 10/pack
₹7,900.00INR / Pkg of 10
Koflo 1/4-24P Unmounted in-line mixer; 1/4"OD, 24 elements, 10/pack WW-04667-14 Mfr # 1/4-24P
COMPARE
6 24 Unmounted in-line mixer; 1/4\"OD, 24 elements, 10/pack
₹13,000.00INR / Pkg of 10
Koflo 3/8-24P Unmounted in-line mixer; 3/8"OD, 24 elements, 10/pack WW-04667-16 Mfr # 3/8-24P
COMPARE
9.5 24 Unmounted in-line mixer; 3/8\"OD, 24 elements, 10/pack
₹9,900.00INR / Pkg of 10
Koflo 1/2-24P Unmounted in-line mixer; 1/2"OD, 24 elements, 10/pack WW-04667-18 Mfr # 1/2-24P
COMPARE
10 24 Unmounted in-line mixer; 1/2\"OD, 24 elements, 10/pack
₹14,100.00INR / Pkg of 10
Showing 1- 8 of 8