This product is now discontinued, View Related Products
Ultratech Ultra Spill Deck Bladder Systems
Hidden containment bladder automatically unfurls to capture spills or leaks
Low profile, 5-3/4" high spill decks conserves valuable floor space and ensures easy transport of drums onto deck. Unique, polyethylene bladder remains rolled up within deck — deploys only if spill occurs and allows an additional 55 gallons of containment.
Compliance: EPA 40 CFR 264.175
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity (Gal)
Availability Pricing
UltraTech 1320 Deck/Pallet Bladder System, 1 Drum/77 gal WW-86452-00 Mfr # 1320
COMPARE
77
Call For Price
UltraTech 2329 Deck/Pallet Bladder System, 2 Drum/99 gal WW-86452-02 Mfr # 2329
COMPARE
99
Call For Price
UltraTech 2330 Deck/Pallet Bladder System, 4 Drum/110 gal WW-86452-04 Mfr # 2330
COMPARE
110