UFM Vortex Flowmeters/Transmitters
A rugged design for aggressive environments
  • Ideal for flow measurement of industrial fluid mixtures and coolants
  • The efficient display is easy to read for process monitoring and unit programming
These flowmeters offer a rugged design and an array of output options at a low cost. With no moving parts and a heavy-duty brass body, the device was designed to operate in aggressive environments. Meters are ideal for measuring process water, water-based machine coolant, or water/glycol coolants and can alert process operators of dangerous flow deviations.
Programming each unit is done through a simple array of buttons on the face of the unit. Through the 3-digit LED, you may select the display units (GPM or LPM), the relay setpoint, and the relay function (normally open or normally closed). Rotate the display for easy viewing within any system arrangement.
Order a multipin cable separately to interface this device with a power supply and input/output controller/recorder.
109,000.00 - ₹244,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max Flow Rate (GPM) Min Flow Rate (GPM) Pipe Size
Availability Pricing
UFM CP-1/4 Flowmeter, Vortex, Heavy-Duty Brass, 1/4", 0.4 to 4.0 gpm WW-32997-30 Mfr # CP-1/4
COMPARE
₹109,000.00INR / Each
UFM CP-1/2 Flowmeter, Vortex, Heavy-Duty Brass, 1/2", 1.2 to 12 gpm WW-32997-32 Mfr # CP-1/2
COMPARE
₹113,000.00INR / Each
UFM CP-1 Flowmeter, Vortex, Heavy-Duty Brass, 1", 5 to 50 gpm WW-32997-36 Mfr # CP-1
COMPARE
₹184,000.00INR / Each
UFM CP-2 Flowmeter, Vortex, Heavy-Duty Brass, 2", 20 to 200 gpm WW-32997-40 Mfr # CP-2
COMPARE
₹244,000.00INR / Each