This product is now discontinued, View Related Products
Two-Sided Spatulas, Polyamide
Strong polyamide spatulas are glass fiber reinforced. Good chemical resistance against solvents; poor resistance to acids or oxidizing agents. Choose either double spatula or spoon/spatula style. Flat spatula end measures 11⁄4" (3.2 cm) L x 3⁄4" (1.9 cm) W. Spoon end measures 11⁄4" (3.2 cm) L and holds 1⁄2 tsp (2.5 mL).
4,200.00 - ₹8,000.00INR / Pkg of 10
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Overall Length (in)
Availability Pricing
Dynalon Two-sided polyamide spatula, double spatula WW-06260-40 Mfr # 391925-0002
COMPARE
7
₹4,200.00INR / Pkg of 10
Dynalon Two-sided polyamide spatula, spoon/spatula WW-06260-42 Mfr # 391925-0003
COMPARE
7
₹8,000.00INR / Pkg of 10