This product is now discontinued, View Related Products
Troemner Class-000 UltraClass Platinum Analytical Calibration Weights
UltraClass Platinum -The weight tolerance is equal to ANSI/ASTM E617 Class 000 and Class E0*. This class is used as a primary standard for calibrating other reference standards and weights where the stability of the environment and careful handling are assured. This class is appropriate for calibrating high-precision analytical balances with a readability as low as 0.0001 mg.
  • UltraClass Platinum tolerance = ASTM Class 000
  • UltraClass weights, 1 g through 5 kg, are made from exclusive Troemner Alloy 8 stainless steel
  • UltraClass Platinum weights come with a positive calibration tolerance gaurantee.
These weights are developed to meet the most demanding calibration needs with the ability to be adjusted. Consistent automated measurement means that Troemner UltraClass weights are the most precise two-piece weights available with weight tolerances that equal or exceed comparable ANSI/ASTM E617 Classes.
Weights are designed so the specifications on surface finish,markings, construction, material density, and magnetic properties become more restrictive as the weight tolerances
decrease in order to enable the weight to maintain its tolerance over time. These weights are designed for your most demanding weighing applications.
Suggested Markets for Analytical Precision Weights include, but are not limited to, pharmaceutical, biotech, nanotechnology, life science, forensic and nuclear industries.
42 PRODUCT OPTIONS
Compareitem specifications
Mass Certification Mass Size (g) Mass Size (mg)
Availability Pricing
Troemner 1 mg, Ultraclass Platinum Mass, Statement of Accuracy WW-01020-01 Mfr # 7037-UCP
COMPARE
Statement of Accuracy 0.001 1
Call For Price
Troemner 2 mg, Ultraclass Platinum Mass, Statement of Accuracy WW-01020-02 Mfr # 7036-UCP
COMPARE
Statement of Accuracy 0.002 2
Call For Price
Troemner 5 mg, Ultraclass Platinum Mass, Statement of Accuracy WW-01020-03 Mfr # 7034-UCP
COMPARE
Statement of Accuracy 0.005 5
Call For Price
Troemner 10 mg, Ultraclass Platinum Mass, Statement of Accuracy WW-01020-04 Mfr # 7033-UCP
COMPARE
Statement of Accuracy 0.01 10
Call For Price
Troemner 20 mg, Ultraclass Platinum Mass, Statement of Accuracy WW-01020-05 Mfr # 7032-UCP
COMPARE
Statement of Accuracy 0.02 20
Call For Price
Troemner 50 mg, Ultraclass Platinum Mass, Statement of Accuracy WW-01020-06 Mfr # 7030-UCP
COMPARE
Statement of Accuracy 0.05 50
Call For Price
Troemner 100 mg, Ultraclass Platinum Mass, Statement of Accuracy WW-01020-07 Mfr # 7029-UCP
COMPARE
Statement of Accuracy 0.1 100
Call For Price
Troemner 200 mg, Ultraclass Platinum Mass, Statement of Accuracy WW-01020-08 Mfr # 7028-UCP
COMPARE
Statement of Accuracy 0.2 200
Call For Price
Troemner 500 mg, Ultraclass Platinum Mass, Statement of Accuracy WW-01020-09 Mfr # 7026-UCP
COMPARE
Statement of Accuracy 0.5 500
Call For Price
Troemner 1g, Ultraclass Platinum Mass, Statement of Accuracy WW-01020-10 Mfr # 7025-UCP
COMPARE
Statement of Accuracy 1 1000
Call For Price
Troemner 2g, Ultraclass Platinum Mass, Statement of Accuracy WW-01020-11 Mfr # 7024-UCP
COMPARE
Statement of Accuracy 2 2000
Call For Price
Troemner 5g, Ultraclass Platinum Mass, Statement of Accuracy WW-01020-12 Mfr # 7022-UCP
COMPARE
Statement of Accuracy 5 5000
Call For Price
Troemner 10g, Ultraclass Platinum Mass, Statement of Accuracy WW-01020-13 Mfr # 7021-UCP
COMPARE
Statement of Accuracy 10 10000
Call For Price
Troemner 20g, Ultraclass Platinum Mass, Statement of Accuracy WW-01020-14 Mfr # 7020-UCP
COMPARE
Statement of Accuracy 20 20000
Call For Price
Troemner 50g, Ultraclass Platinum Mass, Statement of Accuracy WW-01020-15 Mfr # 7018-UCP
COMPARE
Statement of Accuracy 50 50000
Call For Price
Troemner 100g, Ultraclass Platinum Mass, Statement of Accuracy WW-01020-16 Mfr # 7017-UCP
COMPARE
Statement of Accuracy 100 100000
Call For Price
Troemner 200g, Ultraclass Platinum Mass, Statement of Accuracy WW-01020-17 Mfr # 7016-UCP
COMPARE
Statement of Accuracy 200 200000
Call For Price
Troemner 500g, Ultraclass Platinum Mass, Statement of Accuracy WW-01020-18 Mfr # 7014-UCP
COMPARE
Statement of Accuracy 500 500000
Call For Price
Troemner 1kg, Ultraclass Platinum Mass, Statement of Accuracy WW-01020-19 Mfr # 7013-UCP
COMPARE
Statement of Accuracy 1000 1000000
Call For Price
Troemner 2kg, Ultraclass Platinum Mass, Statement of Accuracy WW-01020-20 Mfr # 7012-UCP
COMPARE
Statement of Accuracy 2000 2000000
Call For Price
Showing 1 - 20 of 42