This product is now discontinued, View Related Products

Troemner Class-000 UltraClass Platinum Analytical Calibration Weight Sets

UltraClass Platinum -The weight tolerance is equal to ANSI/ASTM E617 Class 000 and Class E0*. This class is used as a primary standard for calibrating other reference standards and weights where the stability of the environment and careful handling are assured. This class is appropriate for calibrating high-precision analytical balances with a readability as low as 0.0001 mg.
  • UltraClass Platinum tolerance = ASTM Class 000
  • UltraClass weights, 1 g through 5 kg, are made from exclusive Troemner Alloy 8 stainless steel
  • UltraClass Platinum weights come with a positive calibration tolerance gaurantee.
These weights are developed to meet the most demanding calibration needs with the ability to be adjusted. Consistent automated measurement means that Troemner UltraClass weights are the most precise two-piece weights available with weight tolerances that equal or exceed comparable ANSI/ASTM E617 Classes. We guarantee for the life of all Troemner Alloy 8 Stainless Steel Precision Weights that they will maintain extremely low magnetic susceptibility.

Weights are designed so the specifications on surface finish,markings, construction, material density, and magnetic properties become more restrictive as the weight tolerances
decrease in order to enable the weight to maintain its tolerance over time. These weights are designed for your most demanding weighing applications.

Suggested Markets for Analytical Precision Weights include, but are not limited to, pharmaceutical, biotech, nanotechnology, life science, forensic and nuclear industries.
22 PRODUCT OPTIONS
Compareitem specifications
Contents of Set Mass Certification Mass Class
Availability Pricing
Troemner 500mg-1mg, Analytical Prec Ultraclass Platinum Weight Set Statment of Accuracy WW-01022-28 Mfr # 7240-UCP
COMPARE
500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg; polycarbonate case Statement of Accuracy UltraClass Platinum
Call For Price
Troemner 100g-1g, Analytical Ultraclass Platinum Mass Set, Statement of Accuracy WW-01022-29 Mfr # 7224-UCP
COMPARE
100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g; polycarbonate case Statement of Accuracy UltraClass Platinum
Call For Price
Troemner 100g-10 mg, Analytical Ultraclass Platinum Mass Set, Statement of Accuracy WW-01022-31 Mfr # 7226-UCP
COMPARE
100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg; polycarbonate case Statement of Accuracy UltraClass Platinum
Call For Price
Troemner 200g-1 mg, Analytical Ultraclass Platinum Mass Set, Statement of Accuracy WW-01022-32 Mfr # 7223-UCP
COMPARE
200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg; polycarbonate case Statement of Accuracy UltraClass Platinum
Call For Price
Troemner 200g-1 mg, (2), Analytical Ultraclass Platinum Mass Set, Statement of Accuracy WW-01022-33 Mfr # 7221-UCP
COMPARE
(2) 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg; polycarbonate case Statement of Accuracy UltraClass Platinum
Call For Price
Troemner 500g-1 mg, Analytical Ultraclass Platinum Mass Set, Statement of Accuracy WW-01022-34 Mfr # 7219-UCP
COMPARE
500 g, (2) 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg; polycarbonate case Statement of Accuracy UltraClass Platinum
Call For Price
Troemner 1kg-1 mg, Analytical Ultraclass Platinum Mass Set, Statement of Accuracy WW-01022-35 Mfr # 7217-UCP
COMPARE
1 kg, 500 g, (2) 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg; wood case and polycarbonate case Statement of Accuracy UltraClass Platinum
Call For Price
Troemner 50G-10mg, Analytical Ultraclass Platinum Mass Set, Statement of Accuracy WW-01022-36 Mfr # 7231-UCP
COMPARE
50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg; polycarbonate case Statement of Accuracy UltraClass Platinum
Call For Price
Troemner 50g-1g Analytical Ultraclass Platinum Weight Set Statement of Accuracy WW-01022-37 Mfr # 7229-UCP
COMPARE
50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g; polycarbonate case Statement of Accuracy UltraClass Platinum
Call For Price
Troemner 2kg-1g, Analytical Ultraclass Platinum Mass Set, Statement of Accuracy WW-01022-38 Mfr # 7214-UCP
COMPARE
2 kg, 1 kg, 500 g, (2) 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g; wood case and polycarbonate case Statement of Accuracy UltraClass Platinum
Call For Price
Troemner 7240-UCPW 500 mg,-1 mg, Analytical Ultraclass Platinum Weight Set, NVLAP Certificate WW-01022-39 Mfr # 7240-UCPW
COMPARE
500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg; polycarbonate case NIST/NVLAP UltraClass Platinum
Call For Price
Troemner 7224-UCPW 100g-1g Ultraclass Platinum Mass Set, NVLAP Accredited Certificate WW-01022-41 Mfr # 7224-UCPW
COMPARE
100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g; polycarbonate case NIST/NVLAP UltraClass Platinum
Call For Price
Troemner 7226-UCPW 100g-10 mg, Ultraclass Platinum Mass Set, NVLAP Accredited Certificate WW-01022-42 Mfr # 7226-UCPW
COMPARE
100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg; polycarbonate case NIST/NVLAP UltraClass Platinum
Call For Price
Troemner 7223-UCPW 200g-1 mg, Ultraclass Platinum Mass Set, NVLAP Accredited Certificate WW-01022-43 Mfr # 7223-UCPW
COMPARE
200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg; polycarbonate case NIST/NVLAP UltraClass Platinum
Call For Price
Troemner 7221-UCPW 200g-1 mg, (2) Ultraclass Platinum Mass Set, NVLAP Accredited Certificate WW-01022-44 Mfr # 7221-UCPW
COMPARE
(2) 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg; polycarbonate case NIST/NVLAP UltraClass Platinum
Call For Price
Troemner 7219-UCPW 500g-1 mg, Ultraclass Platinum Mass Set, NVLAP Accredited Certificate WW-01022-45 Mfr # 7219-UCPW
COMPARE
500 g, (2) 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg; polycarbonate case NIST/NVLAP UltraClass Platinum
Call For Price
Troemner 7217-UCPW 1kg-1 mg, Ultraclass Platinum Mass Set, NVLAP Accredited Certificate WW-01022-46 Mfr # 7217-UCPW
COMPARE
1 kg, 500 g, (2) 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg; wood case and polycarbonate case NIST/NVLAP UltraClass Platinum
Call For Price
Troemner 7231-UCPW 50g-10 mg, Ultraclass Platinum Mass Set, NVLAP Accredited Certificate WW-01022-47 Mfr # 7231-UCPW
COMPARE
50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg; polycarbonate case NIST/NVLAP UltraClass Platinum
Call For Price
Troemner 7229-UCPW 50g-1g Ultraclass Platinum Mass Set, NVLAP Accredited Certificate WW-01022-48 Mfr # 7229-UCPW
COMPARE
50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g; polycarbonate case NIST/NVLAP UltraClass Platinum
Call For Price
Troemner 7214-UCPW 2kg-1g Ultraclass Platinum Mass Set, NVLAP Accredited Certificate WW-01022-49 Mfr # 7214-UCPW
COMPARE
2 kg, 1 kg, 500 g, (2) 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g; wood case and polycarbonate case NIST/NVLAP UltraClass Platinum
Call For Price
Showing 1 - 20 of 22

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat