This product is now discontinued, View Related Products

Troemner Class-00 UltraClass Gold Analytical Calibration Weight Sets

UltraClass Gold - The weight tolerance is equal to ANSI/ASTM E617 Class 00 and meets or exceeds OIML R 111 Class E1. This class is used as a reference standard for calibrating other reference standards and weights where the stability of the environment and careful handling are assured. This class is appropriate for calibrating high-precision analytical balances with a readability as low as 0.001 mg.
  • UltraClass Glold tolerance = ASTM Class 00
  • Weights include a Statement of Accuracy
  • UltraClass weights, 1 g through 5 kg, are made from exclusive Troemner Alloy 8 stainless steel
These weights are developed to meet the most demanding calibration needs with the ability to be adjusted. Consistent automated measurement means that Troemner UltraClass weights are the most precise two-piece weights available with weight tolerances that equal or exceed comparable ANSI/ASTM E617 Classes.

Weights are designed so the specifications on surface finish,markings, construction, material density, and magnetic properties become more restrictive as the weight tolerances
decrease in order to enable the weight to maintain its tolerance over time. These weights are designed for your most demanding weighing applications.

Suggested Markets for Analytical Precision Weights include, but are not limited to, pharmaceutical, biotech, nanotechnology, life science, forensic and nuclear industries.
22 PRODUCT OPTIONS
Compareitem specifications
Contents of Set Mass Certification Mass Class
Availability Pricing
Troemner 500 Mg-1 mg, Analytical Ultraclass Gold Weight Set Statement of Accuracy WW-01022-51 Mfr # 7240-UCG
COMPARE
500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg; polycarbonate case Statement of Accuracy UltraClass Gold
Call For Price
Troemner 100g-1g Ultraclass Gold Mass Set, Statement of Accuracy WW-01022-52 Mfr # 7224-UCG
COMPARE
100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g; polycarbonate case Statement of Accuracy UltraClass Gold
Call For Price
Troemner 100g-10 mg, Ultraclass Gold Mass Set, Statement of Accuracy WW-01022-53 Mfr # 7226-UCG
COMPARE
100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg; polycarbonate case Statement of Accuracy UltraClass Gold
Call For Price
Troemner 200g-10 mg, Ultraclass Gold Mass Set, Statement of Accuracy WW-01022-54 Mfr # 7223-UCG
COMPARE
200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg; polycarbonate case Statement of Accuracy UltraClass Gold
Call For Price
Troemner 200g-1 mg, (2) Ultraclass Gold Mass Set, Statement of Accuracy WW-01022-55 Mfr # 7221-UCG
COMPARE
(2) 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg; polycarbonate case Statement of Accuracy UltraClass Gold
Call For Price
Troemner 500g-1 mg, Ultraclass Gold Mass Set, Statement of Accuracy WW-01022-56 Mfr # 7219-UCG
COMPARE
500 g, (2) 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg; polycarbonate case Statement of Accuracy UltraClass Gold
Call For Price
Troemner 1kg-1 mg, Ultraclass Gold Mass Set, Statement of Accuracy WW-01022-57 Mfr # 7217-UCG
COMPARE
1 kg, 500 g, (2) 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg; wood case and polycarbonate case Statement of Accuracy UltraClass Gold
Call For Price
Troemner 50g-10 mg, Analytical Ultraclass Gold Weight Set Statement of Accuracy WW-01022-58 Mfr # 7231-UCG
COMPARE
50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg; polycarbonate case Statement of Accuracy UltraClass Gold
Call For Price
Troemner 50g-1g Analytical Ultraclass Gold Weight Set Statement of Accuracy WW-01022-59 Mfr # 7229-UCG
COMPARE
50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g; polycarbonate case Statement of Accuracy UltraClass Gold
Call For Price
Troemner 2kg-1g Analytical Ultraclass Gold Mass Set, Statement of Accuracy WW-01022-60 Mfr # 7214-UCG
COMPARE
2 kg, 1 kg, 500 g, (2) 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g; wood case and polycarbonate case Statement of Accuracy UltraClass Gold
Call For Price
Troemner 7240-UCGW 500 mg,-1 mg, Analytical Ultraclass Gold Weight Set, NVLAP Certificate WW-01022-61 Mfr # 7240-UCGW
COMPARE
500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg; polycarbonate case NIST/NVLAP UltraClass Gold
Call For Price
Troemner 7224-UCGW 100g-1g Ultraclass Gold Mass Set, NVLAP Accredited Certificate WW-01022-62 Mfr # 7224-UCGW
COMPARE
100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g; polycarbonate case NIST/NVLAP UltraClass Gold
Call For Price
Troemner 7226-UCGW 100g-10 mg, Ultraclass Gold Mass Set, NVLAP Accredited Certificate WW-01022-63 Mfr # 7226-UCGW
COMPARE
100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg; polycarbonate case NIST/NVLAP UltraClass Gold
Call For Price
Troemner 7223-UCGW 200g-1 mg, Ultraclass Gold Mass Set, NVLAP Accredited Certificate WW-01022-64 Mfr # 7223-UCGW
COMPARE
200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg; polycarbonate case NIST/NVLAP UltraClass Gold
Call For Price
Troemner 7221-UCGW 200g-1 mg, (2) Ultraclass Gold Mass Set, NVLAP Accredited Certificate WW-01022-66 Mfr # 7221-UCGW
COMPARE
(2) 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg; polycarbonate case NIST/NVLAP UltraClass Gold
Call For Price
Troemner 7219-UCGW 500g-1 mg, Ultraclass Gold Mass Set, NVLAP Accredited Certificate WW-01022-67 Mfr # 7219-UCGW
COMPARE
500 g, (2) 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg; polycarbonate case NIST/NVLAP UltraClass Gold
Call For Price
Troemner 7217-UCGW 1kg-1 mg, Ultraclass Gold Mass Set, NVLAP Accredited Certificate WW-01022-68 Mfr # 7217-UCGW
COMPARE
1 kg, 500 g, (2) 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg; wood case and polycarbonate case NIST/NVLAP UltraClass Gold
Call For Price
Troemner 7231-UCGW 50g-10 mg, Analytical Ultraclass Gold Weight Set, NVLAP Certificate WW-01022-69 Mfr # 7231-UCGW
COMPARE
50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg; polycarbonate case NIST/NVLAP UltraClass Gold
Call For Price
Troemner 7229-UCGW 50g-1g Analytical Ultraclass Gold Weight Set, NVLAP Certificate WW-01022-71 Mfr # 7229-UCGW
COMPARE
50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g; polycarbonate case NIST/NVLAP UltraClass Gold
Call For Price
Troemner 7214-UCGW Ultraclass Gold Mass Set, NVLAP Accredited Certificate; 2kg-1g WW-01022-72 Mfr # 7214-UCGW
COMPARE
2 kg, 1 kg, 500 g, (2) 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g; wood case and polycarbonate case NIST/NVLAP UltraClass Gold
Call For Price
Showing 1 - 20 of 22

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat