Troemner ASTM Class 7 Cast Iron Grip-Handle Calibration Weight with ISO/IEC 17025 Accredited Certificate
ASTM Class 7 cast iron grip-handle weights with ISO/IEC 17025 accredited certificate for audit-ready scale verification
  • Includes ISO/IEC 17025 accredited certificate for documented scale verification records
  • ASTM Class 7 accuracy for checking scales with +/-0.1% accuracy or lower
  • Durable cast iron construction with grip handle for easier lifting, carrying, and placement
  • Gold tone finish identifies metric weights
These ASTM Class 7 cast iron grip-handle weights include an ISO/IEC 17025 accredited certificate for applications that require formal calibration documentation, traceability records, or audit-ready support. Use them for scale verification programs where quality procedures, customer requirements, or regulatory expectations call for an accredited certificate.
Cast iron grip-handle weights larger than 5 kg/10 lb are legal-for-trade in design, making them appropriate for checking weighing devices used in commercial transactions. They are commonly used by weights and measures officials, device installers, and service technicians to test many Class III, IIIL, and IIII scales, as well as scales not marked with a class designation.
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Mass Size (kg)
Availability Pricing
Troemner 9286T 25kg ASTM Class 7 Cast Iron Grip-Handle Calibration Weight with ISO/IEC 17025 Accredited Certificate 0102290 Mfr # 9286T
COMPARE
25
₹72,100.00INR / Each
Troemner 9278-7W 2kg Class F Cast Iron Grip Handle Weight with Traceable Certificate 0102989 Mfr # 9278-7W
COMPARE
2
₹31,100.00INR / Each
Troemner 9284T 20kg ASTM Class 7 Cast Iron Grip-Handle Calibration Weight with ISO/IEC 17025 Accredited Certificate 1196655 Mfr # 9284T
COMPARE
20
₹57,900.00INR / Each
Troemner 9282T 10kg ASTM Class 7 Cast Iron Grip-Handle Calibration Weight with ISO/IEC 17025 Accredited Certificate 1196656 Mfr # 9282T
COMPARE
10
₹44,800.00INR / Each