Tripp Lite Isolation Transformers
Complete line isolation, noise filtering, and surge suppression
  • Faraday Shield offers 100% isolation from input AC line
Isolation transformer design serves as an alternative to installing dedicated circuits and electrical upgrades. Remove EMI/RFI noise, utility switching transients, background spikes, and power, problems generated by other onsite loads, utility grid-switching, and lightning related surges. Surge suppression components offer continuous line filtering of a full range of power line noise in all modes. Transformers feature NEMA 5-15P input and NEMA 5-15R output outlets.
94,300.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Number of Outlets
Availability Pricing
Tripp Lite IS-1000 Isolation Transformer, 1000 W WW-05154-04 Mfr # IS-1000
COMPARE
4
₹94,300.00INR / Each