Porta-Lab Tripod Burner Stands
These single-ring tripod stands feature a cast-iron top and mild steel legs. The top finished in baked hammertone and the legs are plated and removable.
3,800.00 - ₹4,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Height (in) Description
Availability Pricing
Porta-Lab BS-2001 Microburner Stand with Locking Ring, 4" H WW-36310-50 Mfr # BS-2001
COMPARE
4 Microburner Stand with Locking Ring, 4\" H
₹3,800.00INR / Each
Porta-Lab BS-2121 Burner Tripod Stand, 9" H with Locking Taps WW-36310-57 Mfr # BS-2121
COMPARE
9 Burner Tripod Stand, 9\" H with Locking Taps for Accessories
₹4,000.00INR / Each