Monarch Instrument Track-It Temperature and Temperature/Humidity Data Loggers
Optional three-year battery for long-term recording
  • High accuracy and repeatability
  • Multiple user programmable displays
  • Wide temperature range 0 to 185°F (-20C to 85°C)
  • Programmable high and low alarms — perfect for validation
  • Direct USB connection
  • Software provides graphing tools to examine and analyze the data record
Use these high-accuracy data loggers in a multitude of applications. From monitoring food transport conditions to keeping track of temperature and humidity in HVAC, pharmaceutical transports, or lab and museum environments. These compact data loggers will fit anywhere and are simple to use. Each data logger is a battery-powered stand-alone unit that records up to 64,000 sets of temperature and relative humidity data. Choose from a model with a standard battery or long-life battery.
Easily configure units using the included Track-It™ software. Simply plug the logger into an open USB port on your PC and the software automatically identifies the logger; USB interface cable sold separately. Configure the unit to start and stop recording at a predetermined time and date or only when an alarm condition is sensed. The sample storage rate can be set from 1 sample every 2 seconds to 1 sample every 24 hours. A splash-proof protective cover protects the logger when in use.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Temperature Accuracy (° F) Temperature Resolution (° F)
Availability Pricing
Monarch TI-TEMP/DIS/SB Temperature data logger with display and standard battery WW-30003-00 Mfr # 5396-0101
COMPARE
185
Call For Price
Monarch 5396-0102 Temperature data logger with display and long-life battery WW-30003-01 Mfr # 5396-0102
COMPARE
185
Call For Price
Monarch 5396-0201 Temp/RH Data Logger w/ LCD Display; Standard Battery WW-30003-02 Mfr # 5396-0201
COMPARE
185
Call For Price
Monarch 5396-0202 Temp/RH Data Logger w/ LCD Display; Long-Life Battery WW-30003-03 Mfr # 5396-0202
COMPARE
185