Cole-Parmer® Stainless Steel Digital Calipers with Calibration
Versatility and accuracy in a compact design
  • Durable stainless steel construction
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
These stainless steel digital calipers measure inside diameter, outside diameter, and depth. Easily switch from inches to millimeters with the touch of a button. Fine-adjustment thumb roller improves reading speed and position-holding lock screw sets length.
An individually-numbered Traceable® Certificate is provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement.”
11,600.00 - ₹13,200.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Max range (in) Resolution
Availability Pricing
COMPARE
±0.01 in 6 0.0005 in
₹11,600.00INR / Each
COMPARE
±0.001 in 8 0.0005 in
₹13,200.00INR / Each