This product is now discontinued, View Related Products
Traceable® Large-Digit Stopwatches/Chronographs with Calibration
Single-event, time-out/time-in, cumulative-split, and continuous timing
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Shock-resistant ABS plastic case for rugged durability
  • Features AM/PM, date, day, alarm, and clock functions
  • Use as a timer or use to display time of day in hours, minutes, and seconds
These stopwatch/timer/chronographs feature extra-tough construction and time-of-day alarm—ideal for industrial and plant use.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
2 PRODUCT OPTIONS
Compareitem specifications
Accuracy Certifications Timing Capacity
Availability Pricing
COMPARE
±0.001% NIST-traceable 23 hours, 59 minutes, 59 seconds
Call For Price
COMPARE
±0.01% NIST-traceable 23 hours, 59 minutes, 59 seconds