Traceable® High-Temperature Wi-Fi Data Logging Thermometer Compatible with TraceableLIVE® Cloud Service
Stay connected to your critical environments — wherever you go!
  • Connect to your data via smartphone, tablet, or PC using TraceableLIVE wireless capability*
  • View current temperature conditions anywhere—cloud-based interface sends email, text, and push notifications
  • No software required—web, Android™, Apple® iOS data visibility
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
These data logging thermometers with Wi-Fi offer an easy and secure way to connect your smartphone, tablet, or personal computer remotely with your water baths, ovens, incubators, and biological/pharmaceutical lab equipment.
Once setup is complete you can securely monitor current temperatures, control your alarm parameters remotely, view your data logging history, run reports in real time, and give access to other team members. No local software is required to use the cloud-based data interface. Get mobile push notifications, emails, or texts for temperature alarms, connectivity interruptions, or low-battery condition. Up to 672 records can be stored in seven days if the device is set to 15-minute intervals.
Thermometers feature 6" (15.2-cm) L stainless steel probe(s) with 10-ft (3-m) cable. Model 18400-26 includes one probe and model 18400-27 includes two probes.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
44,600.00 - ₹50,900.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Min Temperature (° C) Description
Availability Pricing
COMPARE
300 0 Calibrated High-Temperature Wi-Fi Data Logger Compatible with TraceableLIVE® Cloud Service; 1 Probe
₹44,600.00INR / Each
COMPARE
300 0 Calibrated High-Temperature Wi-Fi Data Logger Compatible with TraceableLIVE® Cloud Service; 2 Probes
₹50,900.00INR / Each