Traceable® Flashing LED Big-Digit Timers with Calibration
Green LED flashes for the last 10 seconds, then flashes red at zero
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Store three countdown times in memory for your most common uses
Large digits permit viewing from across the lab. When counting down, green LED flashes every second for the last ten seconds; red LED flashes when zero is reached and the alarm is sounding. Adjustable alarm volume has three settings: high, low, and off. Timer also counts up showing elapsed time after zero was reached. Features magnetic back, flip-open easel stand, and an opening for a lanyard.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
3,500.00 - ₹6,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Number of Channels Timing Capacity
Availability Pricing
COMPARE
±0.01% 1 99 min 59 seconds
₹3,500.00INR / Each
COMPARE
±0.01% 2 99 hours, 59 minutes, 59 seconds
₹6,000.00INR / Each