Traceable® Expanded Range Dual-Input Thermometers with Calibration
Measure and monitor two readings simultaneously
  • Ideal for a wide range of applications with –40 to 2000°F temperature range
  • Meter features two thermocouple inputs for simultaneous monitoring
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
Meter features an ultra-fast response time with update rate in less than one second. Enhanced features include low battery indicator and ⅝" high 4 digit LCD.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
14,600.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Max Temperature (° F) Description
Availability Pricing
COMPARE
1200 2192 Calibrated Dual-Input Expanded Range Thermocouple Thermometer, Type K; Celsius
₹14,600.00INR / Each