This product is now discontinued, View Related Products
Traceable® Analog Dial Power Controllers with Calibration
Automatic mechanical units turn on and off any piece of equipment
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
Controllers are easy to use — simply set the dial to the present time, pull up/push down pins based on your needs, plug the timer in and plug in the item to be controlled. Manual override switch allows you to turn on your equipment without disturbing the program.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Number of Programs Resolution Timing Capacity
Availability Pricing
COMPARE
96 on/off events per day 15 min 24 hours
Call For Price
COMPARE
12 on/off events per day 2 hr 7 days