Torbal FSB Chuck Clamp Torque Gauge
2.75" LCD display with backlight
  • Six modes and applications
  • Wireless data transmitter
  • 1000Hz data capture
  • Onscreen stats and analytics
  • Onscreen graphing and histogram
High precision chuck clamp torque guage designed for clockwise and counterclockwise testing. Easily connect any bit, grip, or attachment as small as 1/16" that is compatible with conventional chuck clamps. Detachable and ergonomic loadcell handles provide comfort and stability while operating the torque sensor in your hands. Multiple interfaces including wireless, USB, RS232, and anolg l/O allows connection to PC, laptop or tablet and control of ecternal peripherals. Easil;t expand memory and save results for download to PC with microSD memory card.
Call For Price
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Capacity (lb-in) Capacity (N-m) Grads.
Availability Pricing
Torbal FSB2 Digital Torque Meter with Chuck Clamp, 17 lb in capacity WW-25304-76 Mfr # FSB2
COMPARE
17 2 Nm, N.cm, kgf.m, gf.m, lbf.in