Thermo Scientific™ TSX High-Performance Lab Refrigerators
Maintain uniform temperature stability—even with sporadic door openings!
  • Keep samples safe with audible and visual alarms for setpoint security, power loss, door ajar, and over or under temperature conditions
  • ENERGY STAR® certified designed to reduce energy usage without compromising performance
  • Superior V-drive technology adjusts cooling performance to conditions inside and outside the refrigerator
The Thermo Scientific™ TSX High-Performance Lab Refrigerators are designed to protect samples while maintaining sustainability. The innovative V-drive technology runs the compressors at variable speeds to adjust cooling requirements to the conditions in and around the refrigerator. When there are frequent door openings or when samples are added to the refrigerator, the control system detects the activity and increases the drive speed to bring the temperature back to set point quickly. When conditions are stable, the drive runs at a low speed, reducing energy consumption while maintaining a stable temperature. This positive forced air circulation maintains temperature uniformity to protect important samples. This technology along with superior insulation keeps the refrigerator up to 10 times quieter at 52 dB.
Samples are kept safe with easy-to-access controls featuring setpoint security. The key-operated switch allows the set point to be locked with alarm to minimize errors and prevent tampering. Touch keypad with digital indicator also features a graphic thermometer allowing for easy viewing of temperature conditions. Steady green light indicates normal operation, flashing top bar designates overtemperature and flashing bulb indicates under temperature.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Refrigerator Capacity (cu ft) Description
Availability Pricing
Thermo Scientific TSX1205SA Refrigerator, Solid Door, 11.5 cu ft; 120 V, 60 Hz WW-44202-18 Mfr # TSX1205SA
COMPARE
120 11.5 Refrigerator, Solid Door, 11.5 cu ft; 120 V, 60 Hz
Call For Price
Thermo Scientific TSX3005SD Refrigerator, Solid Door, 29.2 cu ft; 208 V, 60 Hz WW-44202-23 Mfr # TSX3005SD
COMPARE
208 29.2 Refrigerator, Solid Door, 29.2 cu ft; 208 V, 60 Hz