This product is now discontinued, View Related Products

Thermo Scientific TSX Enzyme Freezer

Store enzymes securely and sustainably in a –20°C manual-defrost freezer
  • ENERGY STAR® certified to save money and the environment
  • V-drive technology maintains temperature uniformity by detecting usage patterns
  • Whisper quiet with a sound level of 52 dBA
  • Easy mobility with four 2"casters; front two are lockable
  • Enzyme bins included for convenient storage
Safely store temperature sensitive enzymes without the risk of damaging temperature fluctuations associated with auto defrost units.

Designed to support sample protection and sustainability objectives, TSX Series high-performance -20°C manual defrost enzyme freezers feature unique V-drive technology. This facilitates ultimate temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising sample integrity.

Features include easy-to-use microprocessor control with setpoint security and alarms, ergonomic door handle and compatibility with SMART VUE® wireless monitoring systems.

In addition to being ENERGY STAR® certified, natural R290 refrigerants make the TSX Series compliant with the U.S. Environmental Protection Agency’s Significant New Alternatives Policy (SNAP).
Call For Price

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Freezer Capacity (cu ft) Number of Doors Power (VAC)
Availability Pricing
Thermo Scientific TSX3020EA Enzyme Freezer, 30 cu ft; 120V/60 Hz WW-44202-48 Mfr # TSX3020EA
COMPARE
30 1 115
Call For Price
Thermo Scientific TSX3020ED Enzyme Freezer, 30 cu ft; 208V/60 Hz WW-44202-49 Mfr # TSX3020ED
COMPARE
30 1 208 / 230
Call For Price
Thermo Scientific TSX3020EV Enzyme Freezer, 30 cu ft; 230V/50 Hz WW-44202-51 Mfr # TSX3020EV
COMPARE
30 1 230
Call For Price
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat