This product is now discontinued, View Related Products
Thermo Scientific TSG General Purpose Refrigerators
Go green while improving cold storage
  • EPA ENERGY STAR® certified high-efficiency cooling system
  • Green, natural refrigerants offer greater safety
  • Manufacured in a zero-waste-to-landfill facility
  • Whisper-quiet compressor and low heat output fits in any lab setting
  • Temperature high and low, door ajar, and power failure alarms protect critical supplies and samples
Ideal for storing reagents, media, and samples, these general purpose refrigerators are also engineered to be better for the environment. Save electricity and money with adaptive defrost, decreased compressor cycle time, and reduced heat transfer. The natural refrigerant R290 will not deplete the ozone layer or contribute substantially to climate change. Being manufactured in a zero-waste-to-landfill facility ensures at least 99% of the waste produced is reused, recycled, composted, incinerated, or used in energy recovery.
The refrigerator's quiet 51-dBA operation and low heat output allows for installation near workstations for added convenience. Visible and audible alarms when temperature exceeds high or low limits, when the doors are ajar, and when there is a power outage prevent losses of valuable contents. Durable steel construction withstands chemical spills and resists corrosion.
1,319,000.00 - ₹3,032,000.00INR / Each
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
Compareitem specifications
Number of Doors Refrigerator Capacity (cu ft) Description
Availability Pricing
Thermo Scientific TSG General Purpose Refrigerator, 12 cuft, White Exterior, Glass Door; 115 VAC WW-44261-01 Mfr # TSG12RPGA
COMPARE
1 glass 12 General Purpose Refrigerator, 12 cuft, White Exterior, Glass Door; 115 VAC
₹1,319,000.00INR / Each
Thermo Scientific TSG General Purpose Refrigerator, 23 cuft, White Exterior, Glass Door; 115 VAC WW-44261-03 Mfr # TSG25RPGA
COMPARE
1 glass 23 General Purpose Refrigerator, 23 cuft, White Exterior, Glass Door; 115 VAC
₹1,396,000.00INR / Each
Thermo Scientific TSG General Purpose Refrigerator, 23 cuft, White Exterior, Solid Door; 115 VAC WW-44261-05 Mfr # TSG25RPSA
COMPARE
1 solid 23 General Purpose Refrigerator, 23 cuft, White Exterior, Solid Door; 115 VAC
₹1,506,000.00INR / Each
Thermo Scientific TSG General Purpose Refrigerator, 23 cuft, Gray Exterior, Glass Door; 115 VAC WW-44261-07 Mfr # TSG25RSGA
COMPARE
1 glass 23 General Purpose Refrigerator, 23 cuft, Gray Exterior, Glass Door; 115 VAC
₹1,786,000.00INR / Each
Thermo Scientific TSG General Purpose Refrigerator, 23 cuft, Gray Exterior, Solid Door; 115 VAC WW-44261-09 Mfr # TSG25RSSA
COMPARE
1 solid 23 General Purpose Refrigerator, 23 cuft, Gray Exterior, Solid Door; 115 VAC
₹1,751,000.00INR / Each
Thermo Scientific TSG General Purpose Refrigerator, 27 cuft, White Exterior, Glass Door; 115 VAC WW-44261-11 Mfr # TSG30RPGA
COMPARE
1 glass 27 General Purpose Refrigerator, 27 cuft, White Exterior, Glass Door; 115 VAC
₹1,533,000.00INR / Each
Thermo Scientific TSG General Purpose Refrigerator, 27 cuft, White Exterior, Solid Door; 115 VAC WW-44261-13 Mfr # TSG30RPSA
COMPARE
1 solid 27 General Purpose Refrigerator, 27 cuft, White Exterior, Solid Door; 115 VAC
₹1,654,000.00INR / Each
Thermo Scientific TSG General Purpose Refrigerator, 27 cuft, Gray Exterior, Glass Door; 115 VAC WW-44261-15 Mfr # TSG30RSGA
COMPARE
1 glass 27 General Purpose Refrigerator, 27 cuft, Gray Exterior, Glass Door; 115 VAC
₹1,957,000.00INR / Each
Thermo Scientific TSG General Purpose Refrigerator, 27 cuft, Gray Exterior, Solid Door; 115 VAC WW-44261-17 Mfr # TSG30RSSA
COMPARE
1 solid 27 General Purpose Refrigerator, 27 cuft, Gray Exterior, Solid Door; 115 VAC
₹1,919,000.00INR / Each
Thermo Scientific TSG General Purpose Refrigerator, 45 cuft, White Exterior, Sliding Glass Door; 115 VAC WW-44261-19 Mfr # TSG45RPLA
COMPARE
2 sliding glass 45 General Purpose Refrigerator, 45 cuft, White Exterior, Sliding Glass Door; 115 VAC
₹2,092,000.00INR / Each
Thermo Scientific TSG General Purpose Refrigerator, 45 cuft, Gray Exterior, Sliding Glass Door; 115 VAC WW-44261-21 Mfr # TSG45RSLA
COMPARE
2 sliding glass 45 General Purpose Refrigerator, 45 cuft, Gray Exterior, Sliding Glass Door; 115 VAC
₹2,174,000.00INR / Each
Thermo Scientific TSG General Purpose Refrigerator, 49 cuft, White Exterior, Double Glass Doors; 115 VAC WW-44261-23 Mfr # TSG49RPGA
COMPARE
2 glass 49 General Purpose Refrigerator, 49 cuft, White Exterior, Double Glass Doors; 115 VAC
₹1,993,000.00INR / Each
Thermo Scientific TSG General Purpose Refrigerator, 49 cuft, White Exterior, Double Solid Doors; 115 VAC WW-44261-25 Mfr # TSG49RPSA
COMPARE
2 solid 49 General Purpose Refrigerator, 49 cuft, White Exterior, Double Solid Doors; 115 VAC
₹1,955,000.00INR / Each
Thermo Scientific TSG General Purpose Refrigerator, 49 cuft, Gray Exterior, Double Glass Doors; 115 VAC WW-44261-27 Mfr # TSG49RSGA
COMPARE
2 glass 49 General Purpose Refrigerator, 49 cuft, Gray Exterior, Double Glass Doors; 115 VAC
Call For Price
Thermo Scientific TSG General Purpose Refrigerator, 49 cuft, Gray Exterior, Double Solid Doors; 115 VAC WW-44261-29 Mfr # TSG49RSSA
COMPARE
2 solid 49 General Purpose Refrigerator, 49 cuft, Gray Exterior, Double Solid Doors; 115 VAC
₹2,345,000.00INR / Each
Thermo Scientific TSG General Purpose Refrigerator, 72 cuft, White Exterior, Triple Glass Doors; 115 VAC WW-44261-31 Mfr # TSG72RPGA
COMPARE
3 glass 72 General Purpose Refrigerator, 72 cuft, White Exterior, Triple Glass Doors; 115 VAC
₹2,600,000.00INR / Each
Thermo Scientific TSG General Purpose Refrigerator, 72 cuft, Gray Exterior, Triple Glass Doors; 115 VAC WW-44261-33 Mfr # TSG72RSGA
COMPARE
3 glass 72 General Purpose Refrigerator, 72 cuft, Gray Exterior, Triple Glass Doors; 115 VAC
₹3,032,000.00INR / Each
Showing 1- 17 of 17