This product is now discontinued, View Related Products
Thermo Scientific™ TRACE TR-5 (5% Phenyl Methylpolysiloxane) GC Columns
Low bleed provides reproducible and accurate results
Thermo Scientific TRACE GC columns have been tested and found to provide less than half the bleed of competitive capillary columns. Low bleeding means fewer problems with low sensitivity, contamination of the detector, lower temperature limits, and shorter life spans. These columns have been manufactured with improved bonding to be extremely robust, minimizing the risk of damage due to contaminated carrier gases. Improved column-to-column reproducibility guarantees that your column will work with the same high standard as those purchased years from now. Each column goes through rigorous testing and is delivered with a certificate of traceability to guarantee the performance.
TRACE TR-5 Columns contain a 5% Phenyl methylpolysiloxane phase and have a slightly higher polarity than the TR-1 and TR-1MS column, giving it a broad application range with extremely low bleed and a maximum operating temperature of 350°C. These columns are ideal for method development, alcohols, free fatty acids, aromatics, flavors, and low-polarity pesticides.
Call For Price
19 variations of this product are available.
See All Product Options Below
19 PRODUCT OPTIONS
Compareitem specifications
Column Inside Diameter (ID) (mm) Column Length (m) Film Thickness (µm)
Availability Pricing
Thermo Scientific TRACE 260E113P GC Column 7-m length, 0.32-mm ID, 0.25 µm film thickness WW-01852-50 Mfr # 260E113P
COMPARE
0.32 7 0.25
Call For Price
Thermo Scientific TRACE 260E130P GC Column; 15 m L x 0.25 mm ID x 0.25 um thick WW-01852-52 Mfr # 260E130P
COMPARE
0.25 15 0.25
Call For Price
Thermo Scientific TRACE 260E142P GC Column; 30 m L x 0.25 mm ID x 0.25 um thick WW-01852-54 Mfr # 260E142P
COMPARE
0.25 30 0.25
Call For Price
Thermo Scientific TRACE 260E154P GC Column; 60 m L x 0.25 mm ID x 0.25 um thick WW-01852-56 Mfr # 260E154P
COMPARE
0.25 60 0.25
Call For Price
Thermo Scientific TRACE 260E223P GC Column; 30 m L x 0.25 mm ID x 0.50 um thick WW-01852-58 Mfr # 260E223P
COMPARE
0.25 30 0.5
Call For Price
Thermo Scientific TRACE 260E296P GC Column 30-m length, 0.25-mm ID, 1.00 µm film thickness WW-01852-60 Mfr # 260E296P
COMPARE
0.25 30 1
Call For Price
Thermo Scientific TRACE 260E308P GC Column; 60 m L x 0.25 mm ID x 1.0 um thick WW-01852-62 Mfr # 260E308P
COMPARE
0.25 60 1
Call For Price
Thermo Scientific TRACE 260E131P GC Column; 15 m L x 0.32 mm ID x 0.25 um thick WW-01852-64 Mfr # 260E131P
COMPARE
0.32 15 0.25
Call For Price
Thermo Scientific TRACE 260E143P GC Column 30-m length, 0.32-mm ID, 0.25 µm film thickness WW-01852-66 Mfr # 260E143P
COMPARE
0.32 30 0.25
Call For Price
Thermo Scientific TRACE 260E155P GC Column; 60 m L x 0.32 mm ID x 0.25 um thick WW-01852-68 Mfr # 260E155P
COMPARE
0.32 60 0.25
Call For Price
Thermo Scientific TRACE 260E224P GC Column; 30 m L x 0.32 mm ID x 0.50 um thick WW-01852-70 Mfr # 260E224P
COMPARE
0.32 30 0.5
Call For Price
Thermo Scientific TRACE 260E297P GC Column; 30 m L x 0.32 mm ID x 1.0 um thick WW-01852-72 Mfr # 260E297P
COMPARE
0.32 30 1
Call For Price
Thermo Scientific TRACE 260E309P GC Column; 60 m L x 0.32 mm ID x 1.0 um thick WW-01852-74 Mfr # 260E309P
COMPARE
0.32 60 1
Call For Price
Thermo Scientific TRACE 260E225P GC Column; 30 m L x 0.53 mm ID x 0.50 um thick WW-01852-76 Mfr # 260E225P
COMPARE
0.53 30 0.5
Call For Price
Thermo Scientific TRACE 260E286P GC Column; 15 m L x 0.53 mm ID x 1.0 um thick WW-01852-78 Mfr # 260E286P
COMPARE
0.53 15 1
Call For Price
Thermo Scientific TRACE GC Column; 30 m L x 0.53 mm ID x 1.0 um thick WW-01852-80 Mfr # 260E298P
COMPARE
0.53 30 1
Call For Price
Thermo Scientific TRACE 260E334P GC Column; 15 m L x 0.53 mm ID x 1.50 um thick WW-01852-82 Mfr # 260E334P
COMPARE
0.53 15 1.5
Call For Price
Thermo Scientific TRACE 260E336P GC Column; 30 m L x 0.53 mm ID x 1.50 um thick WW-01852-84 Mfr # 260E336P
COMPARE
0.53 30 1.5
Call For Price
Thermo Scientific TRACE 260E470P GC Column; 30 m L x 0.53 mm ID x 5.0 um thick WW-01852-86 Mfr # 260E470P
COMPARE
0.53 30 5
Call For Price
Showing 1- 19 of 19