Thermo Scientific™ SureSTART™ Certified Snap Cap Vials
Certified performance vials are quality tested to deliver the reproducibility you need
  • No more crimping or decapping
  • Compatible with any 11 mm Snap-It or crimp seal
  • 2-mL size is ideal for chromatography applications
These Thermo Scientific™ SureSTART™ 2 mL glass snap vials are Specification Certified to ensure their physical dimensions and autosampler compatibility. Use 11 mm snap caps or 11 mm crimp caps to seal this vial (caps sold separately). Snap caps provide a very tight seal due to their soft polyethylene (PE) material and double snap function. These glass vials offer a secure seal without the hassles of crimping and decapping. The precision-formed crown and snap-on, snap-off cap maximize convenience.
Choose from Level 2 or 3 compliance. Level 2 products have very low metal contents, extractables, and leachables to ensure that they conform to the high standards you need to meet your regulatory requirements. Level 3 products are packed in clean rooms to protect against environmental factors. They contain the lowest levels of metals, extractables, and leachables to ensure consistent, reproducible sensitivity and selectivity for the most challenging analytes.
4,400.00 - ₹4,800.00INR / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Level Tube Color Volume (mL)
Availability Pricing
Thermo Scientific SureSTART™ Certified Snap Cap Autosampler Vials, Clear Glass, Level 3, 2 mL; 100/PK WW-98812-00 Mfr # 6PRV11-1P
COMPARE
3 Clear 2
₹4,800.00INR / Pkg of 100
Thermo Scientific SureSTART™ Certified Snap Cap Autosampler Vials, Amber Glass, Level 2, 2 mL; 100/PK WW-98812-02 Mfr # 6ARV11-2P
COMPARE
2 Amber 2
₹4,400.00INR / Pkg of 100