This product is now discontinued, View Related Products
Thermo Scientific™ National Target™ Screw-Thread Vials
Threaded vials offer a secure closure
  • Manufactured from clear, Type 1 Class A glass and a 10-425 thread finish
These glass screw thread vials are designed specifically for autosamplers that require vials to hang from trays or supports. The wide neck of the vial, and the wider cap profile ensure that vials hang securely.
Vials with write-on patches have graduations at 0.5, 1.0, and 1.5 mL. Caps have a 10-425 thread finish.
5,300.00INR / Pkg of 100
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description Tube Color Volume (mL)
Availability Pricing
Target Screw-Top Autosampler Vials, Clear w/o Caps, 2.0 mL; 100/Pk WW-98813-00 Mfr # C4010-1
COMPARE
Screw-Top Autosampler Vials, Clear w/o Caps, 2.0 mL; 100/Pk Clear 2
₹5,300.00INR / Pkg of 100
Target Screw-Thread Vials, clear with write-on patch WW-98813-01 Mfr # C4010-1W
COMPARE
Screw-Thread Vials, clear with write-on patch Clear 2
Call For Price
Target C4010-2W Screw-Thread Vials, amber with write-on patch WW-98813-02 Mfr # C4010-2W
COMPARE
Screw-Thread Vials, amber with write-on patch Amber 2