Thermo Scientific™ SureSTART™ Level 2 Certified Screw Vial and Cap Kits
Vial kits combine vials and caps with septa for your convenience
  • Certified level 2 vials are ideal for high-throughput analysis
  • 2-mL size is ideal for chromatography applications
  • Marking spot with graduations at 0.5, 1.0, and 1.5-mL
Ensure that your environmental analyses meet government regulations with the Level 2 Thermo Scientific™ SureSTART™ certified short thread screw vials and caps. These convenience kits contain 2 mL glass screw vials and blue, open-top, polypropylene caps with a silicone/PTFE septa. AVCS™ screw caps incorporate Advanced Vial Closure System technology to ensure optimal seal compression when closing a vial.
Level 2 products have very low metal contents and leachables to ensure that they conform to the high standards you need to meet your regulatory requirements. The septa are conditioned to provide a very low level of chromatographic extractables. These products support routine analyses and deliver high quality data with an uninterrupted workflow.
7,500.00 - ₹12,100.00INR / Pkg of 100
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Septum Type Tube Color Volume (mL)
Availability Pricing
Thermo Scientific SureSTART™ AVCS™ Certified Screw Top Vial and Cap Kit, Level 2, Short Thread, Clear Glass, Red Silicone/Ivory PTFE Septa, 2 mL; 100/PK WW-08922-58 Mfr # 6AK80W
COMPARE
Red Silicone/Ivory PTFE Clear 2
₹7,500.00INR / Pkg of 100
Thermo Scientific SureSTART™ AVCS™ Certified Screw Top Vial and Cap Kit, Level 2, Short Thread, Amber Glass, Red Silicone/Ivory PTFE Septa, 2 mL; 100/PK WW-08922-61 Mfr # 6AK82W
COMPARE
Red Silicone/Ivory PTFE Amber 2
₹7,900.00INR / Pkg of 100
Thermo Scientific SureSTART™ AVCS™ Certified Screw Top Vial and Cap Kit, Level 2, Short Thread, Clear Glass, White Silicone/Red PTFE Septa, 2 mL; 100/PK WW-98810-91 Mfr # 6AK92W
COMPARE
White Silicone/Red PTFE Clear 2
₹12,100.00INR / Pkg of 100