Thermo Scientific SAHARA Heated PPO Circulating Baths
Economical alternative to stainless steel baths with better heat resistance than acrylic
  • Control temperatures up to 100°C
  • Two controller options to precisely suit your requirements
These versatile baths provide additional features and flexibility than previous heated baths. Each unit features a removable controller that can be positioned for viewing on all four sides. Intuitive controller with bright digital display allows for easier operation and more precise control. Modify your display by choosing from multiple languages, a temperature resolution of 0.1 or 0.001, and a temperature display in Fahrenheit, Celsius, or Kelvin. High-temperature acoustic and optical alarms protect your samples and equipment from damage. Polyphenylene oxide (PPO) baths allow for heating to 100°C. All models have inlet/outlet connections for external heating applications.
758,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC) Reservoir Capacity (L) Description
Availability Pricing
Thermo Scientific SC150-S21P 21L Heated PPO Bath, Amb +13 to 100C, 115V SKU 1213120 WW-12131-20 Mfr # 1533218
COMPARE
115 21 21L Heated PPO Bath, Amb +13 to 100C, 115V
₹758,000.00INR / Each