Thermo Scientific Owl Self-Recirculating Large-Format Horizontal Electrophoresis Systems
Longer path length for more detailed DNA/RNA analysis
  • Base functions as a built-in three-point leveling platform for flat, uniformly thick gels
  • Pour and run gels right in the gasketed gel tray — no taping or sealing needed
  • UV transparent gel tray lets you view DNA bands after performing runs
Self-recirculating models feature barbed fitting ports for buffer exchange via an external recirculating pump to minimize buffer breakdown and maintain constant pH.
248,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Buffer Solution Required (mL) Gel Thickness (mm) Description
Availability Pricing
Thermo Scientific A1-BP Owl Recirculating Cell, 13 x 25 cm gel size WW-28553-10 Mfr # A1-BP
COMPARE
1600 1.5 Owl Recirculating Cell, 13 x 25 cm gel size
₹248,000.00INR / Each