Thermo Scientific Owl Large Format Non-Recirculating Horizontal Electrophoresis Systems
Longer path length for more detailed DNA/RNA analysis
  • Base functions as a built-in three-point leveling platform for flat, uniformly thick gels
  • Pour and run gels right in the gasketed gel tray — no taping or sealing needed
  • UV transparent gel tray lets you view DNA bands after performing runs
248,000.00INR / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Buffer Solution Required (mL) Gel Thickness (mm) Description
Availability Pricing
Thermo Scientific A2 Non-recirculating cell, 20 x 25 cm gel size WW-28553-20 Mfr # A2-BP
COMPARE
2300 1.5 Non-recirculating cell, 20 x 25 cm gel size
₹248,000.00INR / Each