This product is now discontinued, View Related Products
Thermo Scientific Orion Star T930 Series Ion Titrators
Get fast, reliable, and reproducible automated titrations
  • Provides dedicated ion concentration titrations
  • MKA mode performs automated calibrations
  • Program and save up to 10 user-defined methods
  • Store up to 100 titration data sets with date/time stamp
  • Easily transfer data to printer, computer, or USB drive
  • Minimize downtime with replaceable burettes, tubing, and dispensers
  • Large easy-to-ready touchscreen display enables fast setup
The T900 series automated potentiometric titrators feature a state-of-the-art reagent dispensing system designed to simplify your titrations and offer more reliable, reproducible, and faster results then manual titrations. These compact, rugged titrators are simple to use, let you easily navigate live titration analysis, and are ideal for small bench spaces. You can safely run the titrator unattended until titration is completed.
The automated titrant delivery system, featuring a high-accuracy burette and dispenser, provides consistent results every time. Internal data storage maintains titration history, titrant standardizations, electrode calibrations, and direct measurements, all of which can easily be transferred to an external device.
T930 ion titrators offer dedicated ion concentration titrations including salt in food products, chloride in drinking water and wastewater, ammonia and Total Kjeldajhl Nitrogen (TKN) of wastewater, surfactants in consumer products, and total hardness of drinking water and wastewater. Titration techniques include equivalence point titrations, preset mV endpoint titrations, and multiple known addition (MKA) mode.
1,052,000.00 - ₹1,090,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Memory Size Description
Availability Pricing
Thermo Scientific Orion Star T930 Ion Titrator without Electrode; 100 to 240 VAC WW-94102-35 Mfr # START9300
COMPARE
20 100 data sets Ion Titrator without Electrode; 100 to 240 VAC
₹1,052,000.00INR / Each
Thermo Scientific Orion Star T930 Ion Titrator Chloride/Salt Kit; 100 to -240 VAC WW-94102-36 Mfr # START9301
COMPARE
20 100 data sets Ion Titrator Chloride/Salt Kit; 100 to -240 VAC
₹1,090,000.00INR / Each