Thermo Scientific Orion Star T910 Series pH Titrators
Get fast, reliable, and reproducible automated titrations
  • Provides dedicated acid-base titrations
  • Program and save up to 10 user-defined methods
  • Store up to 100 titration data sets with date/time stamp
  • Easily transfer data to printer, computer, or USB drive
  • Minimize downtime with replaceable burettes, tubing, and dispensers
  • Large easy-to-ready touchscreen display enables fast setup
The T900 series automated potentiometric titrators feature a state-of-the-art reagent dispensing system designed to simplify your titrations and offer more reliable, reproducible, and faster results then manual titrations. These compact, rugged titrators are simple to use, let you easily navigate live titration analysis, and are ideal for small bench spaces. You can safely run the titrator unattended until titration is completed.
The automated titrant delivery system, featuring a high-accuracy burette and dispenser, provides consistent results every time. Internal data storage maintains titration history, titrant standardizations, electrode calibrations, and direct measurements, all of which can easily be transferred to an external device.
T910 pH Titrator is specially designed for acid-based titrations including titratable acidity of juices and wines, acidity of food products, alkalinity of water, acidity and alkalinity of consumer products, total acid number (TAN), and total base number (TBN). Titration techniques include equivalence point titrations and preset pH or mV endpoint titrations for versatile sample analysis.
818,000.00 - ₹899,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Description
Availability Pricing
Thermo Scientific Orion Star T910 pH Titrator without Electrode; 100 to 240 VAC WW-94102-00 Mfr # START9100
COMPARE
20 pH Titrator without Electrode; 100 to 240 V
₹818,000.00INR / Each
Thermo Scientific Orion Star T910 pH Titrator with ROSS Ultra Electrode and ATC Probe; 100 to 240 VAC WW-94102-01 Mfr # START9101
COMPARE
20 pH Titrator with ROSS Ultra electrode & ATC probe; 100 to 240 V
₹892,000.00INR / Each
Thermo Scientific Orion Star T910 pH Titrator with ROSS Sure-Flow electrode and ATC probe; 100 to 240 VAC WW-94102-02 Mfr # START9102
COMPARE
20 pH Titrator with ROSS Sure-Flow electrode and ATC probe; 100 to 240 V
₹899,000.00INR / Each