Thermo™ Scientific™ Nunc™ TripleFlask™ Treated Cell Culture Flasks
Get three flasks in one!
  • Three parallel growth Nunclon™ Delta surfaces provide a total culture area of 500 cm2!
These unique flasks offer three times the yield of standard 175 cm2 flasks in the same space, making them ideal for intermediate scale-up production. Harvest cells using protein digesting enzymes such as trypsin, collagenase, and pronase. The design of this flask prevents mechanical scraping. Total culture area is achieved by way of the three parallel layers and a straight neck design that works effectively within automation systems. TripleFlask receives the Nunclon™ Delta treatment, a proprietary cell culture surface treatment that offers maximum adhesion for a broad range of cell types.
171,000.00INR / Case of 4
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Cap Type Culture Area (cm²) Neck Type
Availability Pricing
Thermo Scientific Nunc TripleFlask™ Cell Culture Flasks, Sterile, Treated, Filter Cap, 200 mL; 32/Cs WW-01931-10 Mfr # 132913
COMPARE
Filter 500 Straight
₹171,000.00INR / Case of 4