Thermo Scientific™ Nunc™ Sterile Cell Culture Petri Dishes with Nunclon™ Delta Surface Treatment
Nunclon™ Delta surface treatment promotes maximum adhesion for a broad range of cell types
  • Top and bottom stacking rings for easier handling and more stable stacking
  • Sterile and non-pyrogenic
  • Constructed of durable polystyrene
  • Lid features vented for air flow
Perform a variety of applications in dishes with the Nunclon™ Delta surface treatment that promotes maximum adhesion for a broad range of cell types. Petri dishes are vented for air flow. Gridded model options feature a grid bottom specially designed for cloning applications and determining plating efficiencies. All dishes are sterilized by gamma irradiation.
106,000.00 - ₹132,000.00INR / Case of 400
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Sterile Surface Treatment Description
Availability Pricing
Thermo Scientific Nunc Cell Culture Petri Dishes, Nunclon™ Delta Treated, Sterile, Non-Gridded, 60 x 15 mm; 400/Cs WW-01930-32 Mfr # 150288
COMPARE
Yes Nunclon™ Delta Cell Culture Petri Dishes, Nunclon™ Delta Treated, Sterile, Non-Gridded, 60 x 15 mm; 400/Cs
₹106,000.00INR / Case of 400
Thermo Scientific Nunc Cell Culture Petri Dishes, Nunclon™ Delta Treated, Sterile, Gridded, 60 x 15 mm; 400/Cs WW-01930-34 Mfr # 169558
COMPARE
Yes Nunclon™ Delta Cell Culture Petri Dishes, Nunclon™ Delta Treated, Sterile, Gridded, 60 x 15 mm; 400/Cs
₹132,000.00INR / Case of 400