Thermo Scientific™ Nunc™ Sterile Cell Culture Flasks with Nunclon™ Delta Surface Treatment
Nunclon™ Delta surface treatment provides optimal cell attachment and growth
  • Virgin polystyrene provides excellent optical quality
  • Sterilized by gamma radiation
  • Large-bore necks—easy pipette and cell scraper access
Achieve maximum adhesion for a broad range of cell types with Thermo Scientific™ Nunc™ Cell Culture Treated Flasks. These flasks receive the proprietary Nunclon™ Delta surface treatment to ensure consistent cell growth.
Vented caps promote sufficient gas exchange while filtered caps allow constant airflow and minimize chances of contamination.
61,400.00 - ₹104,000.00INR / Case of 160
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Cap Type Culture Area (cm²) Neck Type
Availability Pricing
Thermo Scientific Nunc Cell Culture Flask, Polystyrene, Sterile, Treated, Angled Neck, Vent Cap, 25 cm2; 160/Cs WW-01930-41 Mfr # 163371
COMPARE
Vented 25 Angled
₹104,000.00INR / Case of 160
Thermo Scientific Nunc Cell Culture Flask, Polystyrene, Sterile, Treated, Straight Neck, Vent Cap, 80 cm2; 50/Cs WW-01930-42 Mfr # 153732
COMPARE
Vented 80 Straight
₹61,400.00INR / Case of 50
Thermo Scientific Nunc Cell Culture Flask, Polystyrene, Sterile, Treated, Straight Neck, Filter Cap, 80 cm2; 50/Cs WW-01932-42 Mfr # 178905
COMPARE
Filter 80 Straight
₹64,200.00INR / Case of 50
Thermo Scientific Nunc Cell Culture Flask, Polystyrene, Sterile, Treated, Straight Neck, Filter Cap, 175 cm2; 32/Cs WW-01932-44 Mfr # 178883
COMPARE
Filter 175 Straight
₹76,900.00INR / Case of 32