Thermo Scientific™ Nunc™ Sterile Non-Treated Cell Culture Multidishes
Non-treated surface offers low cell attachment
  • Certified nonpyrogenic and sterile
  • Excellent optical properties
  • Raised well rims to lower the risk of cross-contamination
  • Lids feature notched corners for good fit and easy orientation
These sterile polystyrene tissue culture dishes are used for unique cell growth applications that do not require the Nunclon Delta surface. Non-treated surface offers low cell attachment. Clear, uniform design provides excellent optical quality, making it a suitable choice for microscopy.
73,500.00 - ₹100,000.00INR / Case of 120
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Max Working Well Volume (mL) Number of Wells Sterile
Availability Pricing
Thermo Scientific Nunc Cell Culture Multidishes, Non-Treated, Polystyrene, Sterile, 4-Well, 1 mL; 120/Cs WW-01928-60 Mfr # 179820
COMPARE
1 4 Yes
₹100,000.00INR / Case of 120
Thermo Scientific Nunc Cell Culture Multidishes, Non-Treated, Polystyrene, Sterile, 6-Well, 3 mL; 75/Cs WW-01928-62 Mfr # 150239
COMPARE
3 6 Yes
₹73,500.00INR / Case of 75
Thermo Scientific Nunc Cell Culture Multidishes, Non-Treated, Polystyrene, Sterile, 12-Well, 2 mL; 75/Cs WW-01928-64 Mfr # 150200
COMPARE
2 12 Yes
₹84,900.00INR / Case of 75
Thermo Scientific Nunc Cell Culture Multidishes, Non-Treated, Polystyrene, Sterile, 24-Well, 1 mL; 75/Cs WW-01928-66 Mfr # 144530
COMPARE
1 24 Yes
₹92,800.00INR / Case of 75