Thermo Scientific™ Nunc™ Lab-Tek™ Sterile Petri Dishes, Polystyrene
Get better cultures
  • Ideal for fungi, bacteria, and other microorganisms
  • Sterilized by gamma irradiation
These sterile and disposable polystyrene (PS) dishes are ideal for culturing fungi, bacteria, and other microorganisms. Dishes work well in robotic dispensers due to flatness and uniform height. Sterilized by gamma irradiation.
49,100.00 - ₹236,000.00INR / Case of 320
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Lid (Included) Well Diameter (mm) Well Shape
Availability Pricing
Thermo Scientific Nunc Lab-Tek™ Petri Dishes, Sterile, Polystyrene, Round, Vented, 90 x 15 mm; 320/Cs WW-14007-01 Mfr # 263991
COMPARE
Yes, Vented 90 Round
₹49,100.00INR / Case of 320
Thermo Scientific Nunc Lab-Tek™ Petri Dishes, Sterile, Polystyrene, Round, Vented, 140 x 20 mm; 80/Cs WW-14007-05 Mfr # 249964
COMPARE
Yes, Vented 140 Round
₹51,500.00INR / Case of 80
Thermo Scientific Nunc Lab-Tek™ Petri Dishes, Sterile, Polystyrene, Square, Vented, 100 x 15 mm; 500/Cs WW-14007-10 Mfr # 4021
COMPARE
Yes, Vented 100 Square
₹236,000.00INR / Case of 500
Thermo Scientific Nunc Lab-Tek™ Petri Dishes, Sterile, Polystyrene, Round, Non-Vented, 150 x 25 mm; 72/Cs WW-14007-14 Mfr # 4014-12
COMPARE
Yes, Non-Vented 150 Round
₹63,300.00INR / Case of 72