Thermo Scientific™ Nunc™ 96-Well Surface Binding Assay Microplates, Polystyrene
Surfaces with different degrees of hydrophilicity—ideal for solid phase immunoassays (ELISA)
  • Engineered for easy/reproducible passive absorption
  • Certified homogeneity of binding
  • Clear polystyrene has excellent optical properties making it ideal for optical measurements
  • Plates are optimized for robotics, plate readers and liquid handlers
Surface binding assay microplate wells facilitate surface adsorption of proteins, antibodies, and other biomolecules. The surface adsorption aids in applications for the basis for quantitative assays such as ELISA.
Choose from treated or untreated plates. Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment. Untreated surfaces are ideal for adding coatings in your lab. MaxiSorp surface treatment is hydrophilic and optimized for binding immunoglobulin but also binds a wide range of other proteins.
48,400.00 - ₹87,200.00INR / Case of 60
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Surface Treatment Well Shape Well Volume (mL)
Availability Pricing
Thermo Scientific Nunc Surface Binding Assay Microplates, 96-Well, Polystyrene, Non-Treated, Flat, 0.4 mL; 60/Cs WW-01928-02 Mfr # 269620
COMPARE
Non-Treated Flat 0.4
₹48,400.00INR / Case of 60
Thermo Scientific Nunc Surface Binding Assay Microplates, 96-Well, Polystyrene, MaxiSorp® Treated, Round, 0.3 mL; 60/Cs WW-01928-08 Mfr # 449824
COMPARE
MaxiSorp Round 0.3
₹87,200.00INR / Case of 60