Thermo Scientific™ Nalgene™ Class B Polypropylene Volumetric Flask
Won’t leak upon inverting
  • Individually hand calibrated volumetric flask
  • Permanent color graduation line and oval marking spot for ease in reading fluid levels
  • No meniscus to confuse readings
  • Autoclavable
Lighter in weight than glass, Thermo Scientific™ Nalgene™ Class B polypropylene (PP) volumetric flasks with closure offer excellent chemical resistance and are calibrated “To Contain” and “To Deliver” at 20°C (68°F). These volumetric flasks meet ASTM Standard E288 and ISO Standard 384 for consistent quality and dependability.
Flasks have polypropylene screw closures and are leakproof.
9,300.00 - ₹18,800.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL)
Availability Pricing
Thermo Scientific Nalgene Class B Volumetric Flask, Polypropylene, 50 mL WW-06108-10 Mfr # 4000-0050
COMPARE
50
₹9,300.00INR / Each
Thermo Scientific Nalgene Class B Volumetric Flask, Polypropylene, 100 mL WW-06108-20 Mfr # 4000-0100
COMPARE
100
₹9,900.00INR / Each
Thermo Scientific Nalgene Class B Volumetric Flask, Polypropylene, 200 mL WW-06108-30 Mfr # 4000-0200
COMPARE
200
₹11,100.00INR / Each
Thermo Scientific Nalgene Class B Volumetric Flask, Polypropylene, 250 mL WW-06108-40 Mfr # 4000-0250
COMPARE
250
₹12,300.00INR / Each
Thermo Scientific Nalgene Class B Volumetric Flask, Polypropylene, 500 mL WW-06108-50 Mfr # 4000-0500
COMPARE
500
₹14,800.00INR / Each
Thermo Scientific Nalgene Class B Volumetric Flask, Polypropylene, 1000 mL WW-06108-60 Mfr # 4000-1000
COMPARE
1000
₹18,800.00INR / Each