Thermo Scientific Nalgene Rectangular Carboys with Handle, PP
Maintain sample integerity
  • No dead space
  • Leakproof for better handling
  • Easy trasport via steel handle
  • Withstand temperatures to 248°F (120°C)
  • Autoclavable
Space-saving, leakproof polypropylene (PP) carboys feature liter/gallon graduations and built-in shoulder loops with steel handles. Carboys with quick-action spigots accept 5⁄8" ID tubing and come with adapters for 1⁄4" to 1⁄2" ID tubing, integrally molded threaded boss, and two PTFE O-rings for additional versatility. Withstand temperatures to 248°>F (120°>C).
52,300.00 - ₹83,300.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (L) Cap / Stopper Size Material
Availability Pricing
Thermo Scientific Nalgene 2212-0020 rectangular polypropylene carboy with handle, 9 L WW-06082-60 Mfr # 2212-0020
COMPARE
9 100 mm Polypropylene
₹52,300.00INR / Each
Thermo Scientific Nalgene 2212-0050 rectangular polypropylene carboy with handle, 20 L WW-06082-70 Mfr # 2212-0050
COMPARE
20 100 mm Polypropylene
₹75,400.00INR / Each
Thermo Scientific Nalgene 2321-0020 rectangular polypropylene carboy with spigot, 9 L WW-06082-80 Mfr # 2321-0020
COMPARE
9 100 mm Polypropylene
₹61,400.00INR / Each
Thermo Scientific Nalgene 2321-0050 rectangular polypropylene carboy with spigot, 20 L WW-06082-90 Mfr # 2321-0050
COMPARE
20 100 mm Polypropylene
₹83,300.00INR / Each